Product Details

SNP ID
rs201571084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37201847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTCTTGATGATACTTTTTG[C/T]ATCTCTGTTGTGTAGCCAGTCATCA
Phenotype
MIM: 612828
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEBPZ PubMed Links

Gene Details

Gene
CEBPZ
Gene Name
CCAAT/enhancer binding protein zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005760.2 3112 Missense Mutation ACA,GCA T1028A NP_005751.2
Gene
CEBPZOS
Gene Name
CEBPZ opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322373.1 3112 Intron NP_001309302.1
NM_001322374.1 3112 Intron NP_001309303.1
NM_001322375.1 3112 Intron NP_001309304.1
NM_001322376.1 3112 Intron NP_001309305.1
XM_017003106.1 3112 Intron XP_016858595.1
XM_017003107.1 3112 Intron XP_016858596.1
XM_017003108.1 3112 Intron XP_016858597.1
XM_017003109.1 3112 Intron XP_016858598.1

View Full Product Details