Product Details

SNP ID
rs202058287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130729259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCAAGCAGCCAGCAACACTTC[C/G]TTGGGCCTGGGGGACCTCAGGGTGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR148 PubMed Links

Gene Details

Gene
GPR148
Gene Name
G protein-coupled receptor 148
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207364.2 190 Silent Mutation TCC,TCG S36S NP_997247.2
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 190 Intron NP_001264335.1
XM_011511670.2 190 Intron XP_011509972.1
XM_017004732.1 190 Intron XP_016860221.1
XM_017004733.1 190 Intron XP_016860222.1
XM_017004734.1 190 Intron XP_016860223.1
XM_017004735.1 190 Intron XP_016860224.1

View Full Product Details