Product Details

SNP ID
hCV191099721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196296068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCCCAGAAACAGCGAGAAGCCA[C/T]GCTAAAAAATCCAAGAATGAAGAAT
Phenotype
MIM: 123695
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCYT1A PubMed Links

Gene Details

Gene
PCYT1A
Gene Name
phosphate cytidylyltransferase 1, choline, alpha
There are no transcripts associated with this gene.

Gene
TCTEX1D2
Gene Name
Tctex1 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details