Product Details

SNP ID
rs72481822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:36993550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGGCTCTTCTGGCGCCAAAAT[A/G]TCGTTCGTGGCAGGGGTTATTCGGC
Phenotype
MIM: 607911 MIM: 120436
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPM2AIP1 PubMed Links

Gene Details

Gene
EPM2AIP1
Gene Name
EPM2A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014805.3 201 Intron NP_055620.1
Gene
MLH1
Gene Name
mutL homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000249.3 201 Missense Mutation ATA,ATG I1M NP_000240.1
NM_001167617.1 201 Intron NP_001161089.1
NM_001167618.1 201 Intron NP_001161090.1
NM_001167619.1 201 Intron NP_001161091.1
NM_001258271.1 201 Missense Mutation ATA,ATG I1M NP_001245200.1
NM_001258273.1 201 UTR 5 NP_001245202.1
NM_001258274.1 201 Intron NP_001245203.1
XM_005265161.1 201 Missense Mutation ATA,ATG I1M XP_005265218.1
XM_005265163.1 201 Intron XP_005265220.1
XM_005265164.1 201 UTR 5 XP_005265221.1
XM_005265166.1 201 Intron XP_005265223.1
XM_011533727.1 201 Intron XP_011532029.1
XM_017006449.1 201 Intron XP_016861938.1
XM_017006450.1 201 UTR 5 XP_016861939.1
XM_017006451.1 201 UTR 5 XP_016861940.1

View Full Product Details