Product Details

SNP ID
rs199530636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136151593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTTCCTTTACTGCTTTGTCTT[G/T]TTTAAGAATACCTGGCTTGGTTTTA
Phenotype
MIM: 614802 MIM: 604944
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MSL2 PubMed Links

Gene Details

Gene
MSL2
Gene Name
male-specific lethal 2 homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145417.1 2021 Missense Mutation AAA,CAA K356Q NP_001138889.1
NM_018133.3 2021 Missense Mutation AAA,CAA K430Q NP_060603.2
XM_005247571.3 2021 Missense Mutation AAA,CAA K356Q XP_005247628.1
XM_006713684.3 2021 Missense Mutation AAA,CAA K356Q XP_006713747.1
XM_011512949.2 2021 Missense Mutation AAA,CAA K356Q XP_011511251.1
Gene
PPP2R3A
Gene Name
protein phosphatase 2 regulatory subunit B''alpha
There are no transcripts associated with this gene.

View Full Product Details