Product Details

SNP ID
rs202015102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113721878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTAGTTCTTCTTTGTCTCAATA[A/G]TCTCAAAGCCAAACTTCCTGTAGAA
Phenotype
MIM: 610834
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA50 PubMed Links

Gene Details

Gene
NAA50
Gene Name
N(alpha)-acetyltransferase 50, NatE catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308445.1 739 Missense Mutation ACT,ATT T130I NP_001295374.1
NM_025146.3 739 Missense Mutation ACT,ATT T131I NP_079422.1

View Full Product Details