Product Details

SNP ID
rs201915067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136256572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTAGTTTCCTGGAGACAGCGT[C/G]GTCACTGGACGAGGCCGAATCAATG
Phenotype
MIM: 232050
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCCB PubMed Links

Gene Details

Gene
PCCB
Gene Name
propionyl-CoA carboxylase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000532.4 372 Silent Mutation GTC,GTG V107V NP_000523.2
NM_001178014.1 372 Silent Mutation GTC,GTG V107V NP_001171485.1
XM_011512873.1 372 Silent Mutation GTC,GTG V107V XP_011511175.1

View Full Product Details