Product Details
- SNP ID
-
rs201868432
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46742493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCGGGTGTACACACCTGGATTCT[C/G]TGCCTTCACGCAGGCCTTTTCCCAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS45
PubMed Links
Gene Details
- Gene
- PRSS45
- Gene Name
- protease, serine 45
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199183.2 |
544 |
Missense Mutation |
CAG,GAG |
Q182E |
NP_954652.2 |
- Gene
- PRSS46
- Gene Name
- protease, serine 46
There are no transcripts associated with this gene.
View Full Product Details