Product Details

SNP ID
rs201989674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120331228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGAAGAACTTTCTGCATTCTG[C/T]GTGCAGCAACACTAGCTTCATCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC58 PubMed Links

Gene Details

Gene
LRRC58
Gene Name
leucine rich repeat containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099678.1 1184 Missense Mutation CAC,CGC H363R NP_001093148.1

View Full Product Details