Product Details

SNP ID
rs202247714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172506856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTGCAAGTTGCTCAGGAATGAA[A/T]GCCCACTCCTTGATGATTCCCAGGA
Phenotype
MIM: 603598
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFSF10 PubMed Links

Gene Details

Gene
TNFSF10
Gene Name
tumor necrosis factor superfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190942.1 605 UTR 3 NP_001177871.1
NM_001190943.1 605 Intron NP_001177872.1
NM_003810.3 605 Missense Mutation CAT,CTT H161L NP_003801.1

View Full Product Details