Product Details

SNP ID
rs201788253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114128883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTATTGAGAACATGGGTCAAGA[A/G]GAAGGGCAGCCAGCAGACAATGAAG
Phenotype
MIM: 126451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRD3 PubMed Links

Gene Details

Gene
DRD3
Gene Name
dopamine receptor D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000796.5 1459 Missense Mutation CTC,TTC L346F NP_000787.2
NM_001282563.2 1459 Missense Mutation CTC,TTC L346F NP_001269492.1
NM_001290809.1 1459 Missense Mutation CTC,TTC L346F NP_001277738.1
NM_033663.5 1459 Missense Mutation CTC,TTC L313F NP_387512.3
XM_017005829.1 1459 Missense Mutation CTC,TTC L346F XP_016861318.1

View Full Product Details