Product Details

SNP ID
rs201943862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172445268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCCAGAAGTCTGAACACTGCCA[C/G]CCGGTACTTCTTGGACATGATGTTG
Phenotype
MIM: 601898
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GHSR PubMed Links

Gene Details

Gene
GHSR
Gene Name
growth hormone secretagogue receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004122.2 1037 Intron NP_004113.1
NM_198407.2 1037 Missense Mutation CTG,GTG L332V NP_940799.1

View Full Product Details