Product Details

SNP ID
rs201944086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49358049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGATGCCCAAACTGGTTGCAC[C/G]GGAAGCCGAGCACCACCAGGCCCCG
Phenotype
MIM: 138320 MIM: 165390
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPX1 PubMed Links

Gene Details

Gene
GPX1
Gene Name
glutathione peroxidase 1
There are no transcripts associated with this gene.

Gene
RHOA
Gene Name
ras homolog family member A
There are no transcripts associated with this gene.

View Full Product Details