Product Details

SNP ID
rs201002272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180604524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCATGAAAAGCTAGCAGTATC[C/T]CTGTATAGCTCTTCTCTTCCACCAC
Phenotype
MIM: 613798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC39 PubMed Links

Gene Details

Gene
CCDC39
Gene Name
coiled-coil domain containing 39
There are no transcripts associated with this gene.

Gene
TTC14
Gene Name
tetratricopeptide repeat domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042601.2 750 Silent Mutation TCC,TCT S206S NP_001036066.1
NM_001288582.1 750 Silent Mutation TCC,TCT S206S NP_001275511.1
NM_133462.3 750 Silent Mutation TCC,TCT S206S NP_597719.1
XM_011512439.2 750 Intron XP_011510741.1
XM_017005739.1 750 Silent Mutation TCC,TCT S206S XP_016861228.1
XM_017005740.1 750 Silent Mutation TCC,TCT S206S XP_016861229.1

View Full Product Details