Product Details
- SNP ID
-
rs201262177
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51861676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTAAGGACAGGCCATGAGGGTT[C/T]GATTTTGTGTAAGAGACATGGCCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IQCF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114750576] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IQCF2
- Gene Name
- IQ motif containing F2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_203424.1 |
48 |
Nonsense Mutation |
CGA,TGA |
R4* |
NP_982248.1 |
View Full Product Details