Product Details
- SNP ID
-
rs201640456
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:169084915 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTCAACCTTGATAACGTCATAC[A/G]TGGCTTATGGACTGGATAGCACTGG
- Phenotype
-
MIM: 165215
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MECOM
PubMed Links
Gene Details
- Gene
- MECOM
- Gene Name
- MDS1 and EVI1 complex locus
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105077.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1115H |
NP_001098547.3 |
NM_001105078.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1050H |
NP_001098548.2 |
NM_001163999.1 |
3415 |
Silent Mutation |
CAC,CAT |
H1042H |
NP_001157471.1 |
NM_001164000.1 |
3415 |
Silent Mutation |
CAC,CAT |
H1041H |
NP_001157472.1 |
NM_001205194.1 |
3415 |
Silent Mutation |
CAC,CAT |
H1050H |
NP_001192123.1 |
NM_004991.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1238H |
NP_004982.2 |
NM_005241.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1050H |
NP_005232.2 |
XM_005247213.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1239H |
XP_005247270.1 |
XM_005247214.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1230H |
XP_005247271.1 |
XM_005247215.3 |
3415 |
Silent Mutation |
CAC,CAT |
H1229H |
XP_005247272.1 |
XM_005247219.2 |
3415 |
Silent Mutation |
CAC,CAT |
H1051H |
XP_005247276.1 |
XM_005247220.2 |
3415 |
Silent Mutation |
CAC,CAT |
H1051H |
XP_005247277.1 |
XM_005247221.2 |
3415 |
Silent Mutation |
CAC,CAT |
H1051H |
XP_005247278.1 |
XM_005247223.2 |
3415 |
Silent Mutation |
CAC,CAT |
H1050H |
XP_005247280.1 |
XM_005247224.3 |
3415 |
Silent Mutation |
CAC,CAT |
H915H |
XP_005247281.1 |
XM_005247225.3 |
3415 |
Silent Mutation |
CAC,CAT |
H914H |
XP_005247282.1 |
XM_005247226.3 |
3415 |
Silent Mutation |
CAC,CAT |
H905H |
XP_005247283.1 |
XM_011512546.2 |
3415 |
Silent Mutation |
CAC,CAT |
H1123H |
XP_011510848.1 |
XM_017005874.1 |
3415 |
Silent Mutation |
CAC,CAT |
H1115H |
XP_016861363.1 |
XM_017005875.1 |
3415 |
Silent Mutation |
CAC,CAT |
H1041H |
XP_016861364.1 |
XM_017005876.1 |
3415 |
Silent Mutation |
CAC,CAT |
H1042H |
XP_016861365.1 |
XM_017005877.1 |
3415 |
Silent Mutation |
CAC,CAT |
H906H |
XP_016861366.1 |
XM_017005878.1 |
3415 |
Silent Mutation |
CAC,CAT |
H717H |
XP_016861367.1 |
View Full Product Details