Product Details

SNP ID
rs201351415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39184228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTTGCCACCCTCTGGGCAGGG[G/T]ACTGGCTGAGGGAGGGGCAGGTTCA
Phenotype
MIM: 609777
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
XIRP1 PubMed Links

Gene Details

Gene
XIRP1
Gene Name
xin actin binding repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198621.2 5022 UTR 3 NP_001185550.1
NM_194293.2 5022 Missense Mutation ACC,CCC T1740P NP_919269.2
XM_005264909.3 5022 Missense Mutation ACC,CCC T1740P XP_005264966.1

View Full Product Details