Product Details

SNP ID
rs201507702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48927744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATCCCGAGGAGTACCAGTTCAC[C/T]TGCTTGACCTACAAGGAATCTGAGG
Phenotype
MIM: 605615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARIH2 PubMed Links

Gene Details

Gene
ARIH2
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317333.1 502 Silent Mutation ACC,ACT T62T NP_001304262.1
NM_001317334.1 502 Intron NP_001304263.1
NM_006321.3 502 Silent Mutation ACC,ACT T62T NP_006312.1
XM_011533265.1 502 Silent Mutation ACC,ACT T62T XP_011531567.1
XM_011533266.1 502 Silent Mutation ACC,ACT T62T XP_011531568.1
XM_011533267.1 502 Silent Mutation ACC,ACT T62T XP_011531569.1
XM_011533268.1 502 Silent Mutation ACC,ACT T62T XP_011531570.1
XM_011533270.1 502 UTR 5 XP_011531572.1
XM_011533271.1 502 Intron XP_011531573.1
XM_011533273.1 502 UTR 5 XP_011531575.1
XM_011533275.2 502 UTR 5 XP_011531577.1
XM_011533276.1 502 Intron XP_011531578.1
XM_017005531.1 502 Silent Mutation ACC,ACT T62T XP_016861020.1
XM_017005532.1 502 Silent Mutation ACC,ACT T62T XP_016861021.1
XM_017005533.1 502 UTR 5 XP_016861022.1
XM_017005534.1 502 Intron XP_016861023.1
XM_017005535.1 502 Intron XP_016861024.1
XM_017005536.1 502 UTR 5 XP_016861025.1
XM_017005537.1 502 UTR 5 XP_016861026.1
XM_017005538.1 502 UTR 5 XP_016861027.1
XM_017005539.1 502 Intron XP_016861028.1
XM_017005540.1 502 UTR 5 XP_016861029.1
XM_017005541.1 502 UTR 5 XP_016861030.1
Gene
ARIH2OS
Gene Name
ariadne homolog 2 opposite strand
There are no transcripts associated with this gene.

View Full Product Details