Product Details

SNP ID
rs200766815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:154122306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCCCCGGAGTACAGGGCTGCCT[C/T]TCCTCGACTTCGACGGCCCAAGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF26 PubMed Links

Gene Details

Gene
ARHGEF26
Gene Name
Rho guanine nucleotide exchange factor 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251962.1 525 Missense Mutation TCT,TTT S105F NP_001238891.1
NM_001251963.1 525 Missense Mutation TCT,TTT S105F NP_001238892.1
NM_015595.3 525 Missense Mutation TCT,TTT S105F NP_056410.3
XM_011512672.1 525 Missense Mutation TCT,TTT S105F XP_011510974.1
XM_017006160.1 525 Intron XP_016861649.1
Gene
ARHGEF26-AS1
Gene Name
ARHGEF26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details