Product Details

SNP ID
rs201717880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186538709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTCATGTCATTACTCCACAATG[C/T]GGCGGAAAGACTGGACAGCTGGGGA
Phenotype
MIM: 123730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYGS PubMed Links

Gene Details

Gene
CRYGS
Gene Name
crystallin gamma S
There are no transcripts associated with this gene.

Gene
TBCCD1
Gene Name
TBCC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details