Product Details
- SNP ID
-
rs201718003
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:44499975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCCTTACATTCATAAGGTTTCT[A/C]CCCAGTATGGGTTCTCTGGTGGTCG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF852
PubMed Links
Gene Details
- Gene
- ZNF852
- Gene Name
- zinc finger protein 852
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287349.1 |
967 |
Nonsense Mutation |
GAG,TAG |
E268* |
NP_001274278.1 |
View Full Product Details