Product Details

SNP ID
rs201080097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119432159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATAGAGCTGATAGAAGACGACA[C/T]TGCCCTCAATAAGGATGAGCAGATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM39A PubMed Links

Gene Details

Gene
TMEM39A
Gene Name
transmembrane protein 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018266.2 1554 Missense Mutation AAT,AGT N430S NP_060736.1
XM_005247578.1 1554 UTR 3 XP_005247635.1
XM_006713687.1 1554 Missense Mutation AAT,AGT N430S XP_006713750.1
XM_011512958.2 1554 Intron XP_011511260.1
XM_017006788.1 1554 Missense Mutation AAT,AGT N402S XP_016862277.1

View Full Product Details