Product Details

SNP ID
rs201496629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48466993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAACCTGCTCCTAGCCTTCCTGC[A/G]GCGCCAGCCACAGCCCTGGTGCCTG
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272065.1 1474 Intron NP_001258994.1
NM_001272066.1 1474 Intron NP_001258995.1
NM_001272067.1 1474 Intron NP_001258996.1
NM_001272068.1 1474 Intron NP_001258997.1
NM_001272082.1 1474 Intron NP_001259011.1
NM_001272083.1 1474 Intron NP_001259012.1
NM_016479.4 1474 Intron NP_057563.3
XM_005265205.1 1474 Intron XP_005265262.1
XM_006713188.2 1474 Intron XP_006713251.1
XM_011533796.1 1474 Intron XP_011532098.1
Gene
TREX1
Gene Name
three prime repair exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007248.3 1474 Missense Mutation CAG,CGG Q103R NP_009179.2
NM_016381.5 1474 Missense Mutation CAG,CGG Q168R NP_057465.1
NM_033629.4 1474 Missense Mutation CAG,CGG Q113R NP_338599.1

View Full Product Details