Product Details

SNP ID
rs201151319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186613167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGTCCCTCGTCCTGCTCCTTT[G/T]TCTTGCTCAGCTCTGGGGCTGCCAC
Phenotype
MIM: 138680
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AHSG PubMed Links

Gene Details

Gene
AHSG
Gene Name
alpha 2-HS glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001622.2 240 Missense Mutation TGT,TTT C9F NP_001613.2
XM_017005840.1 240 Missense Mutation TGT,TTT C9F XP_016861329.1

View Full Product Details