Product Details

SNP ID
rs200741663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42908117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGCCCTGTACAGGGATGTGAT[A/G]CTGGAGAATTATGGGGCTGTGGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF662 PubMed Links

Gene Details

Gene
ZNF662
Gene Name
zinc finger protein 662
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134656.1 372 Missense Mutation ATA,ATG I62M NP_001128128.1
NM_207404.3 372 Missense Mutation ATA,ATG I1M NP_997287.2

View Full Product Details