Product Details

SNP ID
rs201344304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127197153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTCCAGCCACTGTCCCCAGGT[A/G]GGATGCTGGACTCCAGCCAGCTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf56 PubMed Links

Gene Details

Gene
C3orf56
Gene Name
chromosome 3 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007534.2 708 Silent Mutation GTA,GTG V156V NP_001007535.1

View Full Product Details