Product Details

SNP ID
rs201569855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48621503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAGCGCCGCCGCCCCTGCAGT[A/G]TCTTGCGGCAGATCATCAGCATCGT
Phenotype
MIM: 610068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC26A6 PubMed Links

Gene Details

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
There are no transcripts associated with this gene.

Gene
TMEM89
Gene Name
transmembrane protein 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008269.1 254 Missense Mutation ACA,ATA T85I NP_001008270.1

View Full Product Details