Product Details

SNP ID
rs199837808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8753001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACGCCGTGGATGGCTGGGAGCA[A/G]CTCCTCTGGCTGGAGCTGCGATGGC
Phenotype
MIM: 601253 MIM: 167055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAV3 PubMed Links

Gene Details

Gene
CAV3
Gene Name
caveolin 3
There are no transcripts associated with this gene.

Gene
OXTR
Gene Name
oxytocin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000916.3 1744 Silent Mutation AGC,AGT S382S NP_000907.2
XM_011533762.2 1744 Silent Mutation AGC,AGT S382S XP_011532064.1
Gene
SSUH2
Gene Name
ssu-2 homolog (C. elegans)
There are no transcripts associated with this gene.

View Full Product Details