Product Details

SNP ID
rs200008035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44865053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCTCGCAGGCCTTCCTGGGCT[A/G]TGTGCTGTATGGAGAGTGCCAGGAG
Phenotype
MIM: 600585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR564 PubMed Links

Gene Details

Gene
MIR564
Gene Name
microRNA 564
There are no transcripts associated with this gene.

Gene
TGM4
Gene Name
transglutaminase 4
There are no transcripts associated with this gene.

Gene
TMEM42
Gene Name
transmembrane protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144638.2 369 Missense Mutation TAT,TGT Y118C NP_653239.1

View Full Product Details