Product Details

SNP ID
rs199939644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49807824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGGCGAAAGGCACTACGAGGC[C/T]GTGGCCCACTCACCACCTTATACAG
Phenotype
MIM: 191325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDHR4 PubMed Links

Gene Details

Gene
CDHR4
Gene Name
cadherin related family member 4
There are no transcripts associated with this gene.

Gene
FAM212A
Gene Name
family with sequence similarity 212 member A
There are no transcripts associated with this gene.

Gene
MIR5193
Gene Name
microRNA 5193
There are no transcripts associated with this gene.

Gene
UBA7
Gene Name
ubiquitin like modifier activating enzyme 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003335.2 2626 Missense Mutation CAG,CGG Q876R NP_003326.2
XM_005265430.2 2626 Missense Mutation CAG,CGG Q851R XP_005265487.1
XM_006713321.2 2626 Missense Mutation CAG,CGG Q869R XP_006713384.1
XM_011534070.1 2626 Missense Mutation CAG,CGG Q824R XP_011532372.1
XM_011534071.1 2626 Missense Mutation CAG,CGG Q774R XP_011532373.1
XM_011534072.1 2626 Missense Mutation CAG,CGG Q660R XP_011532374.1

View Full Product Details