Product Details

SNP ID
rs200492126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138262565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTCTAGGTCAGTGATCTTCAAC[C/G]TTGGCTGTGTACTGGACTCACCTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARMC8 PubMed Links

Gene Details

Gene
ARMC8
Gene Name
armadillo repeat containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267041.2 1005 Intron NP_001253970.1
NM_001267042.2 1005 Intron NP_001253971.1
NM_001282342.1 1005 Intron NP_001269271.1
NM_014154.4 1005 Intron NP_054873.2
NM_015396.5 1005 Intron NP_056211.2
NM_213654.3 1005 Intron NP_998819.1
XM_006713564.3 1005 Intron XP_006713627.1
XM_006713565.2 1005 Intron XP_006713628.1
XM_006713566.3 1005 Intron XP_006713629.1
XM_006713567.3 1005 Intron XP_006713630.1
XM_011512632.1 1005 Intron XP_011510934.1
XM_011512633.1 1005 Intron XP_011510935.1
XM_017006095.1 1005 Intron XP_016861584.1
XM_017006096.1 1005 Intron XP_016861585.1
XM_017006097.1 1005 Intron XP_016861586.1
XM_017006098.1 1005 Intron XP_016861587.1
XM_017006099.1 1005 Intron XP_016861588.1
XM_017006100.1 1005 Intron XP_016861589.1
XM_017006101.1 1005 Intron XP_016861590.1
Gene
NME9
Gene Name
NME/NM23 family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178130.3 1005 Missense Mutation ACG,AGG T256R NP_835231.1
XM_005247429.3 1005 Intron XP_005247486.1
XM_006713615.3 1005 Intron XP_006713678.1
XM_011512774.2 1005 Intron XP_011511076.1
XM_011512777.2 1005 Intron XP_011511079.1
XM_017006306.1 1005 Intron XP_016861795.1
XM_017006307.1 1005 Intron XP_016861796.1
XM_017006308.1 1005 Intron XP_016861797.1
XM_017006309.1 1005 Intron XP_016861798.1
XM_017006310.1 1005 Intron XP_016861799.1
XM_017006311.1 1005 Intron XP_016861800.1
XM_017006312.1 1005 Intron XP_016861801.1
XM_017006313.1 1005 Intron XP_016861802.1
XM_017006314.1 1005 Intron XP_016861803.1
XM_017006315.1 1005 Intron XP_016861804.1
XM_017006316.1 1005 Intron XP_016861805.1
XM_017006317.1 1005 Intron XP_016861806.1
XM_017006318.1 1005 Intron XP_016861807.1
XM_017006319.1 1005 Intron XP_016861808.1

View Full Product Details