Product Details

SNP ID
rs199687668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38550597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCGCCTGCTGACGGAAGAGGAA[C/G]GAGGCATGCTTCAAAGAGCGTTGCA
Phenotype
MIM: 600163
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCN5A PubMed Links

Gene Details

Gene
SCN5A
Gene Name
sodium voltage-gated channel alpha subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000335.4 5915 Silent Mutation TCC,TCG S1924S NP_000326.2
NM_001099404.1 5915 Silent Mutation TCC,TCG S1925S NP_001092874.1
NM_001099405.1 5915 Silent Mutation TCC,TCG S1907S NP_001092875.1
NM_001160160.1 5915 Silent Mutation TCC,TCG S1892S NP_001153632.1
NM_001160161.1 5915 Silent Mutation TCC,TCG S1871S NP_001153633.1
NM_198056.2 5915 Silent Mutation TCC,TCG S1925S NP_932173.1
XM_011533991.2 5915 Silent Mutation TCC,TCG S1924S XP_011532293.1
XM_017007017.1 5915 Silent Mutation TCC,TCG S1871S XP_016862506.1

View Full Product Details