Product Details

SNP ID
rs200562529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42908814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAGCATTTCCATTTCCCAAAC[C/T]GGCTCTGATTTCCCAGCTGGAGCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF662 PubMed Links

Gene Details

Gene
ZNF662
Gene Name
zinc finger protein 662
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134656.1 422 Missense Mutation CCG,CTG P79L NP_001128128.1
NM_207404.3 422 Missense Mutation CCG,CTG P19L NP_997287.2

View Full Product Details