Product Details

SNP ID
rs200461678
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.3:187698974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGAAGAGGATGACCACATGGG[C/G]AGACTGCCAGGTGTGCCAGCACCAG
Phenotype
MIM: 609138
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LOC100131635 PubMed Links

Gene Details

Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

Gene
RTP2
Gene Name
receptor transporter protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004312.2 776 Missense Mutation CCC,GCC P68A NP_001004312.2
XM_017006301.1 776 Missense Mutation CCC,GCC P68A XP_016861790.1
XM_017006302.1 776 Missense Mutation CCC,GCC P68A XP_016861791.1

View Full Product Details