Product Details

SNP ID
rs200566629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52249221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGAGCTACAGGAGGATGATGT[A/G]GTTGTCATGGCAACTGATGGACTCT
Phenotype
MIM: 608979 MIM: 611059
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929054 PubMed Links

Gene Details

Gene
LOC101929054
Gene Name
uncharacterized LOC101929054
There are no transcripts associated with this gene.

Gene
PPM1M
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122870.2 1055 Silent Mutation GTA,GTG V166V NP_001116342.1
NM_144641.3 1055 Silent Mutation GTA,GTG V378V NP_653242.3
XM_005264879.2 1055 Silent Mutation GTA,GTG V378V XP_005264936.1
XM_005264880.3 1055 Silent Mutation GTA,GTG V293V XP_005264937.1
XM_005264881.1 1055 Silent Mutation GTA,GTG V217V XP_005264938.1
Gene
WDR82
Gene Name
WD repeat domain 82
There are no transcripts associated with this gene.

View Full Product Details