Product Details
- SNP ID
-
rs200344304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:44500000 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCAGTATGGGTTCTCTGGTGGTC[A/G]AAGAGTTTGGAGCTCTGATTAAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF852
PubMed Links
Gene Details
- Gene
- ZNF852
- Gene Name
- zinc finger protein 852
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287349.1 |
942 |
Silent Mutation |
TTC,TTT |
F259F |
NP_001274278.1 |
View Full Product Details