Product Details

SNP ID
rs199851074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114128800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGGGTTGAGGGCGCTATTCAC[A/G]TAGCCCAGCCATGTCGTGGCACTGT
Phenotype
MIM: 126451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRD3 PubMed Links

Gene Details

Gene
DRD3
Gene Name
dopamine receptor D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000796.5 1542 Silent Mutation TAC,TAT Y373Y NP_000787.2
NM_001282563.2 1542 Silent Mutation TAC,TAT Y373Y NP_001269492.1
NM_001290809.1 1542 Silent Mutation TAC,TAT Y373Y NP_001277738.1
NM_033663.5 1542 Silent Mutation TAC,TAT Y340Y NP_387512.3
XM_017005829.1 1542 Silent Mutation TAC,TAT Y373Y XP_016861318.1

View Full Product Details