Product Details

SNP ID
rs200586913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139344162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCGCTACCTTGCTCTTTCGAG[A/G]TGGGGCTGCCACGCCGCCGGTTCAG
Phenotype
MIM: 605810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS22 PubMed Links

Gene Details

Gene
MRPS22
Gene Name
mitochondrial ribosomal protein S22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020191.2 143 Missense Mutation ATG,GTG M46V NP_064576.1
XM_005247640.2 143 Missense Mutation ATG,GTG M46V XP_005247697.1
XM_006713703.3 143 Missense Mutation ATG,GTG M46V XP_006713766.1
XM_011512995.2 143 Intron XP_011511297.1
XM_011512996.2 143 Intron XP_011511298.1

View Full Product Details