Product Details

SNP ID
rs200639211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183446171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCAAGATCACGCAAAAACCTCT[A/G]CCAAGACAGAACGTGCATCAGCTCA
Phenotype
MIM: 615914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDKN2AIP PubMed Links

Gene Details

Gene
CDKN2AIP
Gene Name
CDKN2A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317343.1 663 UTR 3 NP_001304272.1
NM_017632.3 663 Missense Mutation ACC,GCC T163A NP_060102.1

View Full Product Details