Product Details

SNP ID
rs200755080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165079786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTTCAACAATTTCTTCTAGGC[G/T]TGAAATAGTTCTGCAAGTAATCAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM192 PubMed Links

Gene Details

Gene
TMEM192
Gene Name
transmembrane protein 192
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100389.1 839 Missense Mutation AGC,CGC S230R NP_001093859.1
XM_011531717.2 839 UTR 3 XP_011530019.1
XM_011531718.2 839 Missense Mutation AGC,CGC S185R XP_011530020.1

View Full Product Details