Product Details
- SNP ID
-
rs200047571
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:849760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCATCTTGGCGTGCTGCTCGTA[C/T]GGCTGCCCCGCAGCCTATGGGTGAC
- Phenotype
-
MIM: 602052
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAK
PubMed Links
Gene Details
- Gene
- GAK
- Gene Name
- cyclin G associated kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318134.1 |
4124 |
Silent Mutation |
CCA,CCG |
P1204P |
NP_001305063.1 |
NM_005255.3 |
4124 |
Silent Mutation |
CCA,CCG |
P1283P |
NP_005246.2 |
XM_005272268.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1272P |
XP_005272325.1 |
XM_005272270.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1208P |
XP_005272327.1 |
XM_011513425.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1327P |
XP_011511727.1 |
XM_011513426.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1316P |
XP_011511728.1 |
XM_011513427.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1297P |
XP_011511729.1 |
XM_011513428.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1278P |
XP_011511730.1 |
XM_011513429.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1265P |
XP_011511731.1 |
XM_011513430.1 |
4124 |
Silent Mutation |
CCA,CCG |
P1248P |
XP_011511732.1 |
XM_011513431.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1244P |
XP_011511733.1 |
XM_011513432.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1239P |
XP_011511734.1 |
XM_011513434.2 |
4124 |
Silent Mutation |
CCA,CCG |
P1191P |
XP_011511736.1 |
XM_017007991.1 |
4124 |
Silent Mutation |
CCA,CCG |
P1253P |
XP_016863480.1 |
XM_017007992.1 |
4124 |
Silent Mutation |
CCA,CCG |
P1218P |
XP_016863481.1 |
XM_017007993.1 |
4124 |
Silent Mutation |
CCA,CCG |
P1191P |
XP_016863482.1 |
XM_017007994.1 |
4124 |
Intron |
|
|
XP_016863483.1 |
XM_017007995.1 |
4124 |
Silent Mutation |
CCA,CCG |
P1103P |
XP_016863484.1 |
View Full Product Details