Product Details

SNP ID
rs200968539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69998443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAGAACCCAGCCAACTATGAA[A/G]TTCCTTGTCTTTGCCTTCATCTTGG
Phenotype
MIM: 184470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STATH PubMed Links

Gene Details

Gene
STATH
Gene Name
statherin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009181.1 116 Silent Mutation AAA,AAG K2K NP_001009181.1
NM_003154.2 116 Silent Mutation AAA,AAG K2K NP_003145.1

View Full Product Details