Product Details

SNP ID
rs200678675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144646747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTTCTTTGAAGGAGATGCTAA[G/T]TTTGGGGAAAGAAACGAAGGGAGCG
Phenotype
MIM: 606178
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HHIP PubMed Links

Gene Details

Gene
HHIP
Gene Name
hedgehog interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022475.2 621 Missense Mutation AAG,AAT K24N NP_071920.1
XM_005263178.4 621 Missense Mutation AAG,AAT K24N XP_005263235.1
XM_006714288.3 621 Missense Mutation AAG,AAT K24N XP_006714351.1
Gene
HHIP-AS1
Gene Name
HHIP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details