Product Details

SNP ID
rs200761715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:52024115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCACTGGAGGAACTGGGTAGGC[A/G]GGTGGTGCTGACCATCACAGATCCA
Phenotype
MIM: 600900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC66 PubMed Links

Gene Details

Gene
LRRC66
Gene Name
leucine rich repeat containing 66
There are no transcripts associated with this gene.

Gene
SGCB
Gene Name
sarcoglycan beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000232.4 859 Missense Mutation CGC,TGC R267C NP_000223.1

View Full Product Details