Product Details

SNP ID
rs200242644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:674228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGATCACCTTGATGAGCGGAGAG[A/G]CCTGCACCGGTGGCACCATCTTGTC
Phenotype
MIM: 601519 MIM: 160782 MIM: 180072
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5I PubMed Links

Gene Details

Gene
ATP5I
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007100.3 111 Missense Mutation GCC,GTC A7V NP_009031.1
Gene
MFSD7
Gene Name
major facilitator superfamily domain containing 7
There are no transcripts associated with this gene.

Gene
MYL5
Gene Name
myosin light chain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002477.1 111 Intron NP_002468.1
XM_006713886.2 111 Missense Mutation ACC,GCC T5A XP_006713949.1
XM_017008245.1 111 Intron XP_016863734.1
XM_017008246.1 111 Intron XP_016863735.1
XM_017008247.1 111 Intron XP_016863736.1
XM_017008248.1 111 Intron XP_016863737.1
Gene
PDE6B
Gene Name
phosphodiesterase 6B
There are no transcripts associated with this gene.

View Full Product Details