Product Details

SNP ID
rs200551526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39500188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTGGATCTTGAAGACTAAACTT[C/T]GGAATTTCACCAGAAGGAGCATATG
Phenotype
MIM: 603370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGDH PubMed Links

Gene Details

Gene
UGDH
Gene Name
UDP-glucose 6-dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184700.1 1763 Silent Mutation NP_001171629.1
NM_001184701.1 1763 Silent Mutation NP_001171630.1
NM_003359.3 1763 Silent Mutation NP_003350.1
XM_005262667.2 1763 Silent Mutation XP_005262724.1

View Full Product Details