Product Details

SNP ID
rs200433148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17614787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGACCTTCCAGCAGTACTTTG[C/G]TGGACGAGTTGGAGTCATCTTTCGA
Phenotype
MIM: 170250 MIM: 610311
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LAP3 PubMed Links

Gene Details

Gene
LAP3
Gene Name
leucine aminopeptidase 3
There are no transcripts associated with this gene.

Gene
MED28
Gene Name
mediator complex subunit 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025205.4 160 Missense Mutation CTG,GTG L45V NP_079481.2

View Full Product Details