Product Details

SNP ID
rs199569866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82426116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCCTCGAGATGCCTTGCCATA[A/C]GTGCTCTGTTGGCCTATTTTGAAAA
Phenotype
MIM: 607137
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENOPH1 PubMed Links

Gene Details

Gene
ENOPH1
Gene Name
enolase-phosphatase 1
There are no transcripts associated with this gene.

Gene
HNRNPDL
Gene Name
heterogeneous nuclear ribonucleoprotein D like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207000.1 1741 Silent Mutation ACG,ACT T345T NP_001193929.1
NM_031372.3 1741 Silent Mutation ACG,ACT T402T NP_112740.1

View Full Product Details