Product Details

SNP ID
rs199742995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:52865608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCATTCTATGAAGTGTCTGTC[A/C]GTGAAAATTATAATGATGTCTACAG
Phenotype
MIM: 612404
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RASL11B PubMed Links

Gene Details

Gene
RASL11B
Gene Name
RAS like family 11 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023940.2 730 Missense Mutation AGT,CGT S184R NP_076429.1
Gene
SCFD2
Gene Name
sec1 family domain containing 2
There are no transcripts associated with this gene.

View Full Product Details