Product Details
- SNP ID
-
rs199939366
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:113902385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCTCAGCCTGATTTTTGCTTGG[C/T]TTCTTTTTCTTGGTTTCCTCTTTAT
- Phenotype
-
MIM: 610010
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARSJ
PubMed Links
Gene Details
- Gene
- ARSJ
- Gene Name
- arylsulfatase family member J
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024590.3 |
3172 |
Silent Mutation |
AAA,AAG |
K563K |
NP_078866.3 |
XM_005263212.4 |
3172 |
Silent Mutation |
AAA,AAG |
K563K |
XP_005263269.1 |
XM_011532238.2 |
3172 |
Silent Mutation |
AAA,AAG |
K456K |
XP_011530540.1 |
XM_011532239.2 |
3172 |
Silent Mutation |
AAA,AAG |
K447K |
XP_011530541.1 |
XM_017008592.1 |
3172 |
Silent Mutation |
AAA,AAG |
K563K |
XP_016864081.1 |
XM_017008593.1 |
3172 |
Silent Mutation |
AAA,AAG |
K447K |
XP_016864082.1 |
XM_017008594.1 |
3172 |
Silent Mutation |
AAA,AAG |
K447K |
XP_016864083.1 |
XM_017008595.1 |
3172 |
Silent Mutation |
AAA,AAG |
K447K |
XP_016864084.1 |
XM_017008596.1 |
3172 |
Silent Mutation |
AAA,AAG |
K447K |
XP_016864085.1 |
XM_017008597.1 |
3172 |
Silent Mutation |
AAA,AAG |
K391K |
XP_016864086.1 |
XM_017008598.1 |
3172 |
Silent Mutation |
AAA,AAG |
K391K |
XP_016864087.1 |
XM_017008599.1 |
3172 |
Silent Mutation |
AAA,AAG |
K391K |
XP_016864088.1 |
XM_017008600.1 |
3172 |
Silent Mutation |
AAA,AAG |
K391K |
XP_016864089.1 |
View Full Product Details